Gene Transcripts
- Gene Symbol
- Ppp1r12a
- Entrez Gene ID
- 116670
- Gene Name
- protein phosphatase 1, regulatory subunit 12A
- Gene Aliases
- M110, MBSP, Mypt1
- Chromosome Location
- Chr. 7: 51404971 - 51515382 on Build Rnor_6.0
- UniGene ID
- Rn.162937
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_053890.1 |
NP_446342.1
|
15, 16 |
2225 |
|
XM_006241308.1 |
XP_006241370.1
|
17, 18 |
2668 |
|
XM_006241309.1 |
XP_006241371.1
|
17, 18 |
2668 |
|
XM_006241311.1 |
XP_006241373.1
|
16, 17 |
2554 |
|
XM_006241312.1 |
XP_006241374.1
|
16, 17 |
2500 |
|
XM_006241313.1 |
XP_006241375.1
|
16, 17 |
2488 |
|
XM_006241314.1 |
XP_006241376.1
|
16, 17 |
2554 |
|
XM_006241315.1 |
XP_006241377.1
|
15, 16 |
2374 |
|
XM_006241316.2 |
XP_006241378.1
|
15, 16 |
2700 |
|
GenBank mRNA |
S74907.1 |
- |
15, 16 |
2225 |
|
View Full Product Details