Gene Transcripts
- Gene Symbol
- Wfdc1
- Entrez Gene ID
- 171112
- Gene Name
- WAP four-disulfide core domain 1
- Gene Aliases
- ps20
- Chromosome Location
- Chr. 19: 52313771 - 52332721 on Build Rnor_6.0
- UniGene ID
- Rn.3193
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_133581.1 |
NP_598265.1
|
3, 4 |
440 |
|
XM_006255705.2 |
XP_006255767.1
|
3, 4 |
693 |
|
GenBank mRNA |
AF037272.1 |
- |
3, 4 |
440 |
|
BC063152.1 |
- |
3, 4 |
443 |
6887007 |
FQ213083.1 |
- |
3, 4 |
425 |
|
FQ216345.1 |
- |
3, 4 |
434 |
|
FQ216415.1 |
- |
3, 4 |
435 |
|
FQ219846.1 |
- |
3, 4 |
435 |
|
FQ219958.1 |
- |
3, 4 |
673 |
|
FQ220334.1 |
- |
3, 4 |
672 |
|
FQ229404.1 |
- |
3, 4 |
425 |
|
View Full Product Details