Gene Transcripts

Gene Symbol
LOC257650
Entrez Gene ID
257650
Gene Name
hippyragranin
Gene Aliases
Hgn
Chromosome Location
Chr. 10: 86223336 - 86236884 on Build Rnor_6.0
UniGene ID
Rn.233839
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_147142.1 NP_671483.1 3 226
GenBank mRNA AF260582.1 - 3 226

View Full Product Details