Gene Transcripts

Gene Symbol
Duox1
Entrez Gene ID
266807
Gene Name
dual oxidase 1
Gene Aliases
Chromosome Location
Chr. 3: 114251818 - 114286827 on Build Rnor_6.0
UniGene ID
Rn.162682
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153739.1 NP_714961.1 2 127
XM_006234829.2 XP_006234891.1 3 226
GenBank mRNA AF542180.1 - 2 127

View Full Product Details