Gene Transcripts

Gene Symbol
Kng1l1
Entrez Gene ID
288001
Gene Name
kininogen 1-like 1
Gene Aliases
kininogen
Chromosome Location
Chr. 11: 81421672 - 81444466 on Build Rnor_6.0
UniGene ID
Rn.128333
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009628.1 NP_001009628.1 7 972
GenBank mRNA BC088161.1 - 7 972
M11661.1 - 7 870
M11885.1 - 2 170

View Full Product Details