Gene Transcripts

Gene Symbol
Get4
Entrez Gene ID
288518
Gene Name
golgi to ER traffic protein 4 homolog (S. cerevisiae)
Gene Aliases
Cee, RGD1311660
Chromosome Location
Chr. 12: 17469112 - 17488396 on Build Rnor_6.0
UniGene ID
Rn.102176
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163320.1 NP_001156792.1 5, 6 728
XM_006248927.2 XP_006248989.1 5, 6 2964

View Full Product Details