Gene Transcripts

Gene Symbol
Rbm17
Entrez Gene ID
291295
Gene Name
RNA binding motif protein 17
Gene Aliases
Chromosome Location
Chr. 17: 70586343 - 70603289 on Build Rnor_6.0
UniGene ID
Rn.20694
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013058.1 NP_001013076.1 11 1232
XM_006254205.2 XP_006254267.1 11 1339
GenBank mRNA BC079248.1 - 11 1232
FQ230702.1 - 10 1092

View Full Product Details