Gene Transcripts

Gene Symbol
Nop16
Entrez Gene ID
306768
Gene Name
NOP16 nucleolar protein
Gene Aliases
RGD1305727
Chromosome Location
Chr. 17: 10559811 - 10564728 on Build Rnor_6.0
UniGene ID
Rn.6090
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001047095.1 NP_001040560.1 3 327
GenBank mRNA AB256043.1 - 3 327
AM258958.1 - 3 327
BC158562.1 - 3 331 9088902

View Full Product Details