Gene Transcripts

Gene Symbol
C1QTNF5
Entrez Gene ID
114902
Gene Name
C1q and tumor necrosis factor related protein 5
Gene Aliases
CTRP5
Chromosome Location
Chr. 11: 119338934 - 119346673 on Build GRCh38
UniGene ID
Hs.632102
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278431.1 NP_001265360.1 3 1061
NM_015645.4 NP_056460.1 15 3625
GenBank mRNA AF086482.1 - 1 139
AF329841.1 - 3 1026
AL110261.1 - 2 757
AY358383.1 - 3 1055
BC029485.1 - 3 1062
DB534759.1 - 1 125

Gene Symbol
MFRP
Entrez Gene ID
83552
Gene Name
membrane frizzled-related protein
Gene Aliases
MCOP5, NNO2, RD6
Chromosome Location
Chr. 11: 119338934 - 119346673 on Build GRCh38
UniGene ID
Hs.632102
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031433.3 NP_113621.1 15 3625
GenBank mRNA AJ862823.1 - 15 3607

View Full Product Details