Gene Transcripts
- Gene Symbol
- LINC00202-1
- Entrez Gene ID
- 387644
- Gene Name
- long intergenic non-protein coding RNA 202-1
- Gene Aliases
- C10orf51, LINC00202, NCRNA00202, bB27G4.1
- Chromosome Location
- Chr. 10: 26931206 - 26942001 on Build GRCh38
- UniGene ID
- Hs.529965
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026795.1 |
|
5 |
3576 |
|
GenBank mRNA |
AK097405.1 |
- |
4 |
1743 |
|
AK128027.1 |
- |
4 |
5943 |
|
View Full Product Details