Gene Transcripts

Gene Symbol
HYDIN2
Entrez Gene ID
100288805
Gene Name
HYDIN2, axonemal central pair apparatus protein (pseudogene)
Gene Aliases
HYDINP1
Chromosome Location
Chr. 1: 146547489 - 146822034 on Build GRCh38
UniGene ID
Hs.648712
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_103556.1 11 1533
GenBank mRNA AL833826.1 - 13 1729
AL834340.1 - 8 1023

View Full Product Details