Gene Transcripts
- Gene Symbol
- Wdr72
- Entrez Gene ID
- 546144
- Gene Name
- WD repeat domain 72
- Gene Aliases
- AW558070, D230040E23
- Chromosome Location
- Chr. 9: 74110331 - 74283203 on Build GRCm38
- UniGene ID
- Mm.335289
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK135824.1 |
- |
4 |
680 |
|
BC145956.1 |
- |
4 |
586 |
|
BC145958.1 |
- |
4 |
585 |
|
View Full Product Details