Gene Transcripts

Gene Symbol
Zfp787
Entrez Gene ID
67109
Gene Name
zinc finger protein 787
Gene Aliases
2210018M03Rik, Znf787
Chromosome Location
Chr. 7: 6131489 - 6159564 on Build GRCm38
UniGene ID
Mm.3254
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013012.1 NP_001013030.1 3 978
GenBank mRNA AK077458.1 - 3 946
AK159396.1 - 2 803
BC137753.1 - 3 900 8860772

View Full Product Details