Gene Transcripts

Gene Symbol
Jakmip3
Entrez Gene ID
365380
Gene Name
janus kinase and microtubule interacting protein 3
Gene Aliases
Necc2, RGD1307177
Chromosome Location
Chr. 1: 211293462 - 211421023 on Build Rnor_6.0
UniGene ID
Rn.198526
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163277.1 NP_001156749.1 23 3769
XM_008760004.1 XP_008758226.1 27 4864

View Full Product Details