Gene Transcripts

Gene Symbol
ESYT3
Entrez Gene ID
83850
Gene Name
extended synaptotagmin 3
Gene Aliases
CHR3SYT, E-Syt3, FAM62C
Chromosome Location
Chr. 3: 138434573 - 138479942 on Build GRCh38
UniGene ID
Hs.477711
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001322831.1 NP_001309760.1 1 495
NM_001322834.1 NP_001309763.1 1 495
NM_031913.4 NP_114119.2 1 495
NR_136409.1 1 495
GenBank mRNA AK289530.1 - 1 452
BC037292.1 - 1 487 4837811
DA726011.1 - 1 495
DQ993202.1 - 1 269

View Full Product Details