Gene Transcripts

Gene Symbol
TMEM207
Entrez Gene ID
131920
Gene Name
transmembrane protein 207
Gene Aliases
UNQ846
Chromosome Location
Chr. 3: 190428655 - 190449876 on Build GRCh38
UniGene ID
Hs.335321
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207316.2 NP_997199.1 5 410
GenBank mRNA AY358607.1 - 5 410
BC071780.1 - 5 401 4607584

View Full Product Details