Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_002087.3 | NP_002078.1 | 3 | 450 | |
XM_005257253.1 | XP_005257310.1 | 3 | 495 | ||
GenBank mRNA | AF055008.1 | - | 3 | 256 | |
AK000607.1 | - | 3 | 234 | ||
AK222522.1 | - | 3 | 234 | ||
AK303830.1 | - | 3 | 231 | ||
AY124489.1 | - | 2 | 212 | ||
BC000324.2 | - | 3 | 413 | 2821810 | |
BC010577.2 | - | 2 | 204 | 3457813 | |
BT006844.1 | - | 2 | 194 | ||
DC336362.1 | - | 3 | 450 | ||
M75161.1 | - | 2 | 206 | ||
X62320.1 | - | 3 | 234 |