Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000226.3 | NP_000217.2 | 2, 3 | 793 | |
GenBank mRNA | AB001594.1 | - | 2, 3 | 290 | |
BC121170.2 | - | 3, 4 | 259 | ||
S69510.1 | - | 2, 3 | 790 | ||
Z29074.1 | - | 2, 3 | 793 |