Gene Transcripts
- Gene Symbol
- GLI2
- Entrez Gene ID
- 2736
- Gene Name
- GLI family zinc finger 2
- Gene Aliases
- CJS, HPE9, PHS2, THP1, THP2
- Chromosome Location
- Chr. 2: 120797291 - 120992653 on Build GRCh38
- UniGene ID
- Hs.111867
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_005270.4 |
NP_005261.2
|
10, 11 |
1706 |
|
XM_006712422.2 |
XP_006712485.1
|
10, 11 |
1655 |
|
XM_011510969.2 |
XP_011509271.2
|
10, 11 |
2338 |
|
XM_011510970.2 |
XP_011509272.1
|
10, 11 |
3349 |
|
XM_011510971.2 |
XP_011509273.1
|
10, 11 |
2599 |
|
XM_011510972.2 |
XP_011509274.2
|
10, 11 |
1736 |
|
XM_011510973.2 |
XP_011509275.1
|
9, 10 |
1487 |
|
XM_011510974.2 |
XP_011509276.1
|
9, 10 |
1495 |
|
XM_017003818.1 |
XP_016859307.1
|
10, 11 |
2287 |
|
GenBank mRNA |
AB007295.1 |
- |
7, 8 |
1263 |
|
AB007296.1 |
- |
7, 8 |
1212 |
|
AB007297.1 |
- |
7, 8 |
1263 |
|
AB007298.1 |
- |
7, 8 |
1212 |
|
AB209354.1 |
- |
2, 3 |
283 |
|
BC111410.1 |
- |
10, 11 |
1650 |
|
D14827.1 |
- |
7, 8 |
1212 |
|
D14828.1 |
- |
6, 7 |
1108 |
|
DQ086814.1 |
- |
10, 11 |
1676 |
|
View Full Product Details