Gene Transcripts
- Gene Symbol
- LINC00202-2
- Entrez Gene ID
- 731789
- Gene Name
- long intergenic non-protein coding RNA 202-2
- Gene Aliases
- Chromosome Location
- Chr. 10: 26643108 - 26653454 on Build GRCh38
- UniGene ID
- Hs.178186
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026794.1 |
|
3 |
5319 |
|
GenBank mRNA |
CR749234.1 |
- |
3 |
5259 |
|
- Gene Symbol
- LINC00202-1
- Entrez Gene ID
- 387644
- Gene Name
- long intergenic non-protein coding RNA 202-1
- Gene Aliases
- C10orf51, LINC00202, NCRNA00202, bB27G4.1
- Chromosome Location
- Chr. 10: 26931206 - 26942001 on Build GRCh38
- UniGene ID
- Hs.529965
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026795.1 |
|
3 |
3002 |
|
GenBank mRNA |
AK097405.1 |
- |
2 |
1169 |
|
AK128027.1 |
- |
1, 2 |
5369 |
|
- Gene Symbol
- LINC00264
- Entrez Gene ID
- 645528
- Gene Name
- long intergenic non-protein coding RNA 264
- Gene Aliases
- C10orf50, NCRNA00264, bA128B16.2
- Chromosome Location
- Chr. 10: 26589865 - 26594324 on Build GRCh38
- UniGene ID
- Hs.634511
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026793.1 |
|
1 |
316 |
|
GenBank mRNA |
BC038537.1 |
- |
1 |
316 |
|
View Full Product Details