Gene Transcripts

Gene Symbol
Ar
Entrez Gene ID
11835
Gene Name
androgen receptor
Gene Aliases
AW320017, Tfm
Chromosome Location
Chr. X: 98149750 - 98317147 on Build GRCm38
UniGene ID
Mm.39005
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK133647.1 - 1857
M37890.1 - 3 1796
S56585.1 - 3 1875
X53779.1 - 3 1828
X59411.1 - 3 1827
X59592.1 - 3 1916

View Full Product Details