Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001161419.1 | NP_001154891.1 | 4 | 258 | |
NM_007506.6 | NP_031532.2 | 4 | 251 | ||
GenBank mRNA | AK008191.1 | - | 4 | 247 | |
AK008998.1 | - | 4 | 202 | ||
AK009883.2 | - | 4 | 247 | ||
AK010868.1 | - | 4 | 220 | ||
AK011054.1 | - | 4 | 212 | ||
AK011432.1 | - | 4 | 201 | ||
AK167884.1 | - | 4 | 220 | ||
BC003854.1 | - | 4 | 231 | 3601516 | |
BC094664.1 | - | 188 | 30532297 | ||
BY276643.1 | - | 4 | 259 | ||
L19737.1 | - | 3 | 178 |