Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001136060.2 | NP_001129532.1 | 22 | 2405 | |
NM_001198886.1 | NP_001185815.1 | 22 | 2489 | ||
NM_010075.2 | NP_034205.1 | 22 | 2740 | ||
NM_207282.3 | NP_997165.2 | 22 | 2593 | ||
GenBank mRNA | AF092506.1 | - | 19 | 1753 | |
AF092507.1 | - | 22 | 2748 | ||
AK031903.1 | - | 22 | 2366 | ||
AK158964.1 | - | 22 | 2594 | ||
BC048383.1 | - | 22 | 2256 | 6306267 | |
BC085154.1 | - | 22 | 2489 | 6838230 |