Gene Transcripts

Gene Symbol
Dll4
Entrez Gene ID
54485
Gene Name
delta-like 4 (Drosophila)
Gene Aliases
Delta4
Chromosome Location
Chr. 2: 119325784 - 119335666 on Build GRCm38
UniGene ID
Mm.143719
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019454.3 NP_062327.2 7 1257
GenBank mRNA AB043893.1 - 7 900
AF253469.1 - 7 1212
AF273454.1 - 7 1278
AK004739.1 - 7 1225
AK031699.1 - 7 1226
AK052322.1 - 7 1258
AK165517.1 - 7 1225
AK171717.1 - 7 1224
BC042497.1 - 7 1212 4017786
BC049130.1 - 7 1217 6825525

View Full Product Details