Gene Transcripts

Gene Symbol
1700092M07Rik
Entrez Gene ID
74307
Gene Name
RIKEN cDNA 1700092M07 gene
Gene Aliases
Chromosome Location
Chr. 19: 8740718 - 8741225 on Build GRCm38
UniGene ID
Mm.48791
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001177347.1 NP_001170818.1 2 259
GenBank mRNA AK007043.1 - 2 222
BC048571.1 - 1 343

View Full Product Details