Gene Transcripts
- Gene Symbol
- Olfr395
- Entrez Gene ID
- 259007
- Gene Name
- olfactory receptor 395
- Gene Aliases
- MOR135-10
- Chromosome Location
- Chr. 11: 73906552 - 73907490 on Build GRCm38
- UniGene ID
- Mm.425696
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_147005.1 |
NP_667216.1
|
1 |
72 |
|
View Full Product Details