Gene Transcripts
- Gene Symbol
- FASN
- Entrez Gene ID
- 2194
- Gene Name
- fatty acid synthase
- Gene Aliases
- FAS, OA-519, SDR27X1
- Chromosome Location
- Chr. 17: 82078338 - 82098303 on Build GRCh38
- UniGene ID
- Hs.83190
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_004104.4 |
NP_004095.4
|
42, 43 |
7523 |
|
XM_011523538.2 |
XP_011521840.1
|
42, 43 |
7591 |
|
GenBank mRNA |
AB209988.1 |
- |
42, 43 |
7518 |
|
AY451392.1 |
- |
41, 42 |
7406 |
|
BC007267.1 |
- |
16, 17 |
2665 |
|
BC007909.1 |
- |
11, 12 |
1938 |
|
BC014631.2 |
- |
18, 19 |
3512 |
|
BC014634.2 |
- |
5, 6 |
934 |
|
BC063242.1 |
- |
42, 43 |
7523 |
6172538 |
S80437.1 |
- |
7, 8 |
1277 |
|
U26644.1 |
- |
41, 42 |
7388 |
|
U29344.1 |
- |
42, 43 |
7523 |
|
View Full Product Details