Gene Transcripts

Gene Symbol
FMOD
Entrez Gene ID
2331
Gene Name
fibromodulin
Gene Aliases
FM, SLRR2E
Chromosome Location
Chr. 1: 203340621 - 203351429 on Build GRCh38
UniGene ID
Hs.519168
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002023.4 NP_002014.2 3 2670
NR_103757.1 2 1684
GenBank mRNA AK092899.1 - 4 2131
AK098322.1 - 2 1744
BC035281.1 - 3 2346 5181410
U05291.1 - 2 1313

View Full Product Details