Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_016100.4 | NP_057184.1 | 6 | 871 | |
NM_181527.3 | NP_852668.1 | 6 | 958 | ||
NM_181528.3 | NP_852669.1 | 5 | 725 | ||
GenBank mRNA | AF085355.1 | - | 6 | 834 | |
AL136641.1 | - | 6 | 803 | ||
BC005181.1 | - | 6 | 799 | 3950192 | |
BC008446.1 | - | 6 | 820 | 4104421 |