Gene Transcripts

Gene Symbol
ING2
Entrez Gene ID
3622
Gene Name
inhibitor of growth family member 2
Gene Aliases
ING1L, p33ING2
Chromosome Location
Chr. 4: 183505050 - 183512428 on Build GRCh38
UniGene ID
Hs.107153
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291959.1 NP_001278888.1 2 338
NM_001564.3 NP_001555.1 2 419
XM_011531927.2 XP_011530229.1 2 264
GenBank mRNA AB012853.1 - 2 364
AB196793.1 - 2 213
AF053537.1 - 2 340
AJ006851.1 - 2 273
AK294310.1 - 2 360
AK314000.1 - 2 420
BC030128.2 - 2 399 3941655

View Full Product Details