Gene Transcripts
- Gene Symbol
- LTA4H
- Entrez Gene ID
- 4048
- Gene Name
- leukotriene A4 hydrolase
- Gene Aliases
- Chromosome Location
- Chr. 12: 96000753 - 96043520 on Build GRCh38
- UniGene ID
- Hs.524648
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000895.2 |
NP_000886.1
|
14, 15 |
1524 |
|
NM_001256643.1 |
NP_001243572.1
|
14, 15 |
1455 |
|
NM_001256644.1 |
NP_001243573.1
|
14, 15 |
1455 |
|
NR_132659.1 |
|
14, 15 |
1463 |
|
XM_005268871.1 |
XP_005268928.1
|
14, 15 |
1514 |
|
XM_011538348.1 |
XP_011536650.1
|
14, 15 |
1514 |
|
GenBank mRNA |
AK293630.1 |
- |
14, 15 |
1443 |
|
AK298017.1 |
- |
14, 15 |
1455 |
|
AK301308.1 |
- |
14, 15 |
1333 |
|
AK308184.1 |
- |
14, 15 |
1453 |
|
BC032528.1 |
- |
14, 15 |
1441 |
5502553 |
BX647158.1 |
- |
14, 15 |
1412 |
|
CR457068.1 |
- |
14, 15 |
1382 |
|
J02959.1 |
- |
14, 15 |
1412 |
|
J03459.1 |
- |
14, 15 |
1450 |
|
View Full Product Details