Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_152546.2 | NP_689759.2 | 5 | 375 | |
XM_017009111.1 | XP_016864600.1 | 5 | 419 | ||
GenBank mRNA | AK058015.1 | - | 5 | 375 | |
AY611630.2 | - | 5 | 303 | ||
BC031222.1 | - | 5 | 346 | 5297296 |