Gene Transcripts

Gene Symbol
HSP90B1
Entrez Gene ID
7184
Gene Name
heat shock protein 90 beta family member 1
Gene Aliases
ECGP, GP96, GRP94, HEL-S-125m, HEL35, TRA1
Chromosome Location
Chr. 12: 103930334 - 103947930 on Build GRCh38
UniGene ID
Hs.192374
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003299.2 NP_003290.1 2 311
GenBank mRNA AB209534.1 - 2 207
AJ890084.1 - 2 263
AK025459.1 - 2 263
AK298996.1 - 2 235
AK300521.1 - 2 234
AY040226.1 - 1 66
BC009195.2 - 2 207
BC066656.1 - 2 225 6165138
EU794608.1 - 2 225
FJ460515.1 - 2 225
X15187.1 - 2 234

View Full Product Details