Gene Transcripts
- Gene Symbol
- TOMM7
- Entrez Gene ID
- 54543
- Gene Name
- translocase of outer mitochondrial membrane 7
- Gene Aliases
- TOM7
- Chromosome Location
- Chr. 7: 22812632 - 22822852 on Build GRCh38
- UniGene ID
- Hs.112318
- Species
- Homo sapiens
- Gene Symbol
- C4orf46
- Entrez Gene ID
- 201725
- Gene Name
- chromosome 4 open reading frame 46
- Gene Aliases
- RCDG1
- Chromosome Location
- Chr. 4: 158666675 - 158672255 on Build GRCh38
- UniGene ID
- Hs.380920
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK027539.1 |
- |
|
1643 |
|
View Full Product Details