Gene Transcripts

Gene Symbol
STOML2
Entrez Gene ID
30968
Gene Name
stomatin like 2
Gene Aliases
HSPC108, SLP-2
Chromosome Location
Chr. 9: 35099776 - 35103195 on Build GRCh38
UniGene ID
Hs.3439
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001287031.1 NP_001273960.1 7, 8 764
NM_001287032.1 NP_001273961.1 8, 9 788
NM_001287033.1 NP_001273962.1 7, 8 761
NM_013442.2 NP_038470.1 8, 9 899
GenBank mRNA AF161458.1 - 8, 9 773
AF190167.1 - 8, 9 861
AF282596.1 - 8, 9 832
AK027405.1 - 8, 9 832
AK223102.1 - 8, 9 848
AK225114.1 - 8, 9 832
AK303883.1 - 7, 8 702
BC002442.2 - 8, 9 835 3346384
BC003025.1 - 8, 9 820 2821269
BC010152.2 - 8, 9 832 3534656
BC014990.1 - 8, 9 830 4907745
BU528995.1 - 7, 8 658

View Full Product Details