Gene Transcripts

Gene Symbol
ARPC2
Entrez Gene ID
10109
Gene Name
actin related protein 2/3 complex subunit 2
Gene Aliases
ARC34, PNAS-139, PRO2446, p34-Arc
Chromosome Location
Chr. 2: 218217094 - 218254348 on Build GRCh38
UniGene ID
Hs.529303
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005731.3 NP_005722.1 8, 9 859
NM_152862.2 NP_690601.1 9, 10 954
XM_011510471.1 XP_011508773.1 5, 6 1136
XM_017003113.1 XP_016858602.1 8, 9 828
XM_017003114.1 XP_016858603.1 9, 10 3237
XM_017003115.1 XP_016858604.1 8, 9 818
GenBank mRNA AF006085.1 - 8, 9 869
AF279893.1 - 7, 8 713
AK092520.1 - 5, 6 1916
AK093625.1 - 5, 6 1907
AK290268.1 - 9, 10 853
BC000590.2 - 9, 10 817 3346685
BT006898.1 - 8, 9 785
U50523.1 - 9, 10 869

View Full Product Details