Gene Transcripts

Gene Symbol
TECTA
Entrez Gene ID
7007
Gene Name
tectorin alpha
Gene Aliases
DFNA12, DFNA8, DFNB21
Chromosome Location
Chr. 11: 121102666 - 121190806 on Build GRCh38
UniGene ID
Hs.248162
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005422.2 NP_005413.2 2 86

View Full Product Details