Gene Transcripts
- Gene Symbol
- TRA2B
- Entrez Gene ID
- 6434
- Gene Name
- transformer 2 beta homolog (Drosophila)
- Gene Aliases
- Htra2-beta, PPP1R156, SFRS10, SRFS10, TRA2-BETA, TRAN2B
- Chromosome Location
- Chr. 3: 185914568 - 185938136 on Build GRCh38
- UniGene ID
- Hs.533122
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001243879.1 |
NP_001230808.1
|
6, 7 |
917 |
|
NM_004593.2 |
NP_004584.1
|
7, 8 |
1051 |
|
XM_005247703.1 |
XP_005247760.1
|
8, 9 |
1219 |
|
XM_011513072.2 |
XP_011511374.1
|
8, 9 |
4021 |
|
XM_017007033.1 |
XP_016862522.1
|
7, 8 |
2197 |
|
GenBank mRNA |
AB209208.1 |
- |
8, 9 |
4008 |
|
AK091863.1 |
- |
3, 4 |
1938 |
|
AK094495.1 |
- |
4, 5 |
973 |
|
AK098191.1 |
- |
7, 8 |
838 |
|
AK125610.1 |
- |
4, 5 |
1721 |
|
AK301118.1 |
- |
6, 7 |
770 |
|
AK315632.1 |
- |
7, 8 |
1035 |
|
BC000160.2 |
- |
7, 8 |
918 |
2984771 |
BC000451.2 |
- |
7, 8 |
917 |
2821443 |
BC005898.1 |
- |
7, 8 |
902 |
4046349 |
CR749214.1 |
- |
6, 7 |
3078 |
|
U61267.1 |
- |
7, 8 |
896 |
|
U68063.1 |
- |
7, 8 |
904 |
|
View Full Product Details