Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001277078.1 | NP_001264007.1 | 4 | 1382 | |
NM_004272.4 | NP_004263.1 | 4 | 1382 | ||
XM_017010059.1 | XP_016865548.1 | 4 | 2114 | ||
GenBank mRNA | AF093262.1 | - | 3 | 320 | |
AK223471.1 | - | 4 | 644 | ||
AK312481.1 | - | 4 | 647 | ||
AY189941.1 | - | 4 | 340 | ||
BC015502.1 | - | 4 | 597 | 3904242 | |
BC047438.1 | - | 4 | 1382 | 5294849 | |
BT009846.1 | - | 3 | 320 | ||
Y17829.1 | - | 4 | 395 |