Gene Transcripts

Gene Symbol
ODF1
Entrez Gene ID
4956
Gene Name
outer dense fiber of sperm tails 1
Gene Aliases
CT133, HSPB10, ODF, ODF2, ODF27, ODFP, ODFPG, ODFPGA, ODFPGB, RT7, SODF
Chromosome Location
Chr. 8: 102551620 - 102561017 on Build GRCh38
UniGene ID
Hs.159274
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024410.3 NP_077721.2 1 203
GenBank mRNA AK315276.1 - 1 196
BC104456.1 - 1 174 40030623
BC104457.1 - 1 174 40030627
DB457574.1 - 1 203

View Full Product Details