Gene Transcripts

Gene Symbol
UAP1L1
Entrez Gene ID
91373
Gene Name
UDP-N-acetylglucosamine pyrophosphorylase 1 like 1
Gene Aliases
Chromosome Location
Chr. 9: 137077475 - 137084538 on Build GRCh38
UniGene ID
Hs.142076
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207309.2 NP_997192.2 9 2842
GenBank mRNA AK022632.1 - 4 1727
AK074143.1 - 4 3890
AK095393.1 - 9 2583
AK128529.1 - 7 2677
AL832421.1 - 9 2611
BC106035.1 - 9 2554

View Full Product Details