Gene Transcripts

Gene Symbol
Ptprs
Entrez Gene ID
19280
Gene Name
protein tyrosine phosphatase, receptor type, S
Gene Aliases
AL022616, PTP, PTP-NU3, PTPNU-3, PTPsigma, Ptpt9, R-PTP-S, RPTPsigma
Chromosome Location
Chr. 17: 56412426 - 56476480 on Build GRCm38
UniGene ID
Mm.258771
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001252453.1 NP_001239382.1 12 2221
NM_001252455.1 NP_001239384.1 12 2221
NM_001252456.1 NP_001239385.1 12 2198
NM_011218.2 NP_035348.2 16 3439
GenBank mRNA AK144123.1 - 4 800
BC052462.1 - 12 2147 6834684
BC054077.1 - 12 2115
BC083188.1 - 12 2180 5709264
D28530.1 - 16 3328
D28531.1 - 13 2169
X82288.1 - 15 3248

View Full Product Details