Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_010917.2 | NP_035047.2 | 1 | 299 | |
GenBank mRNA | AK041633.1 | - | 1 | 235 | |
AK049585.1 | - | 1 | 235 | ||
AK084876.1 | - | 1 | 238 | ||
AK144878.1 | - | 1 | 264 | ||
AK166779.1 | - | 1 | 235 | ||
BB840491.1 | - | 1 | 299 | ||
BC052939.1 | - | 1 | 294 | ||
BC131668.1 | - | 1 | 210 | ||
BC131669.1 | - | 1 | 210 | 40110859 | |
X14194.1 | - | 1 | 210 | ||
X14480.1 | - | 1 | 220 |