Gene Transcripts

Gene Symbol
Slc7a3
Entrez Gene ID
11989
Gene Name
solute carrier family 7 (cationic amino acid transporter, y+ system), member 3
Gene Aliases
Atrc3, CAT-3, CAT3, SLC7A1, SLC7A2
Chromosome Location
Chr. X: 101079204 - 101090452 on Build GRCm38
UniGene ID
Mm.5140
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK077593.1 - 6 1117
AK141558.1 - 6 1148
AK165931.1 - 6 1114
BC050195.1 - 6 1093 30010078
U70859.1 - 7 1251

View Full Product Details