Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_007878.2 | NP_031904.1 | 4 | 1284 | |
GenBank mRNA | AK044379.1 | - | 4 | 1253 | |
AK044436.1 | - | 4 | 1275 | ||
BC016086.1 | - | 4 | 1284 | 4501819 | |
BC051421.1 | - | 4 | 1260 | 6492837 |