Gene Transcripts

Gene Symbol
Vwa3a
Entrez Gene ID
233813
Gene Name
von Willebrand factor A domain containing 3A
Gene Aliases
E030013G06Rik
Chromosome Location
Chr. 7: 120735693 - 120805546 on Build GRCm38
UniGene ID
Mm.389939
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177697.3 NP_808365.2 5 794
GenBank mRNA AK086942.1 - 4 392
AK132732.1 - 5 795
AK136898.1 - 4 393

View Full Product Details