Gene Transcripts

Gene Symbol
Hmbox1
Entrez Gene ID
219150
Gene Name
homeobox containing 1
Gene Aliases
AI451877, AI604847, F830020C16Rik
Chromosome Location
Chr. 14: 64811600 - 64949906 on Build GRCm38
UniGene ID
Mm.344074
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177338.5 NP_796312.2 4 845
GenBank mRNA AK052729.1 - 4 855
AK089782.1 - 4 846
AK161592.1 - 4 845
BC002212.1 - 2 249
BC051457.1 - 4 848 6398463

View Full Product Details