Gene Transcripts

Gene Symbol
Msantd4
Entrez Gene ID
78100
Gene Name
Myb/SANT-like DNA-binding domain containing 4 with coiled-coils
Gene Aliases
8430410K20Rik, Mysantd4
Chromosome Location
Chr. 9: 4376413 - 4386890 on Build GRCm38
UniGene ID
Mm.206549
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145609.1 NP_663584.1 2 1055
GenBank mRNA AK032884.1 - 3 1106
AK132023.1 - 3 1091
BC014729.1 - 2 1055 4461077

View Full Product Details