Gene Transcripts

Gene Symbol
2810006K23Rik
Entrez Gene ID
72650
Gene Name
RIKEN cDNA 2810006K23 gene
Gene Aliases
Chromosome Location
Chr. 5: 124328089 - 124341852 on Build GRCm38
UniGene ID
Mm.121658
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028310.3 NP_082586.1 2 1108
GenBank mRNA AK039255.1 - 2 1109

View Full Product Details