Gene Transcripts

Gene Symbol
Tbc1d23
Entrez Gene ID
67581
Gene Name
TBC1 domain family, member 23
Gene Aliases
4930451A13Rik, AU015720, AU043671, AU043778, D030022P07Rik, W51689
Chromosome Location
Chr. 16: 57168858 - 57231532 on Build GRCm38
UniGene ID
Mm.257819
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026254.2 NP_080530.2 17 1972
GenBank mRNA AK015437.1 - 8 804
AK155142.1 - 17 1993
BC019762.1 - 7 710
BC031706.1 - 17 1972 4194266

View Full Product Details